Merck
  • Development and validation of a method for the determination of buprenorphine and norbuprenorphine in breast milk by gas chromatography-mass spectrometry.
Development and validation of a method for the determination of buprenorphine and norbuprenorphine in breast milk by gas chromatography-mass spectrometry.
Biomedical chromatography : BMC (2011-07-02)
Panagiota Nikolaou, Ioannis Papoutsis, Sotirios Athanaselis, Constantinos Pistos, Artemis Dona, Chara Spiliopoulou
ABSTRACT
Buprenorphine (BUP) is used for the maintenance of opioid-addicted pregnant women. Because BUP and its main metabolite nor-BUP are excreted into breast milk, a sensitive and specific GC/MS method has been developed, optimized and validated for their determination in breast milk. BUP-d4 was used as internal standard. The sample preparation includes combination of protein precipitation with solid-phase extraction and derivatization (acetylation). The absolute recovery for both analytes was found to be higher than 87.3%. The limits of detection and quantification were 0.07 and 0.20 µg/L, respectively. The calibration curves were linear within the dynamic range 0.20-20.0 µg/L, with a correlation coefficient higher than 0.996. Intra- and inter-day accuracies were ranged from -7.06 to 4.50 and from -5.88 to 7.00%, respectively, while intra- and inter-day precision were less than 5.7 and 6.1%. The analytes were found to be stable in breast milk at 4 °C for one week, at -20 °C for one month, and after three freeze-thaw cycles. The method can be used for the determination of BUP and nor-BUP in breast milk of BUP-maintained mothers, in order to calculate the amount of drug that could pass to the newborn via breast milk and to avoid toxic consequences of breastfeeding.
Product Description
Supelco
Norbuprenorphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Norbuprenorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®