Merck
  • Heterogenized cobalt oxide catalysts for nitroarene reduction by pyrolysis of molecularly defined complexes.
Heterogenized cobalt oxide catalysts for nitroarene reduction by pyrolysis of molecularly defined complexes.
Nature chemistry (2013-05-23)
Felix A Westerhaus, Rajenahally V Jagadeesh, Gerrit Wienhöfer, Marga-Martina Pohl, Jörg Radnik, Annette-Enrica Surkus, Jabor Rabeah, Kathrin Junge, Henrik Junge, Martin Nielsen, Angelika Brückner, Matthias Beller
ABSTRACT
Molecularly well-defined homogeneous catalysts are known for a wide variety of chemical transformations. The effect of small changes in molecular structure can be studied in detail and used to optimize many processes. However, many industrial processes require heterogeneous catalysts because of their stability, ease of separation and recyclability, but these are more difficult to control on a molecular level. Here, we describe the conversion of homogeneous cobalt complexes into heterogeneous cobalt oxide catalysts via immobilization and pyrolysis on activated carbon. The catalysts thus produced are useful for the industrially important reduction of nitroarenes to anilines. The ligand indirectly controls the selectivity and activity of the recyclable catalyst and catalyst optimization can be performed at the level of the solution-phase precursor before conversion into the active heterogeneous catalyst.
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sigma-Aldrich
Aniline hydrochloride, ≥99%
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol