Merck
  • Feasibility of Tomotherapy-based image-guided radiotherapy to reduce aspiration risk in patients with non-laryngeal and non-pharyngeal head and neck cancer.
Feasibility of Tomotherapy-based image-guided radiotherapy to reduce aspiration risk in patients with non-laryngeal and non-pharyngeal head and neck cancer.
PloS one (2013-03-19)
Nam P Nguyen, Lexie Smith-Raymond, Vincent Vinh-Hung, Paul Vos, Rick Davis, Anand Desai, Thomas Sroka, Dave Abraham, Shane P Krafft, Michelle Stevie, Homayoun Modarresifar, Beng-Hoey Jo, Misty Ceizyk
ABSTRACT
The study aims to assess the feasibility of Tomotherapy-based image-guided radiotherapy (IGRT) to reduce the aspiration risk in patients with non-laryngeal and non-hypopharyngeal cancer. A retrospective review of 48 patients undergoing radiation for non-laryngeal and non-hypopharyngeal head and neck cancers was conducted. All patients had a modified barium swallow (MBS) prior to treatment, which was repeated one month following radiotherapy. Mean middle and inferior pharyngeal dose was recorded and correlated with the MBS results to determine aspiration risk. Mean pharyngeal dose was 23.2 Gy for the whole group. Two patients (4.2%) developed trace aspiration following radiotherapy which resolved with swallowing therapy. At a median follow-up of 19 months (1-48 months), all patients were able to resume normal oral feeding without aspiration. IGRT may reduce the aspiration risk by decreasing the mean pharyngeal dose in the presence of large cervical lymph nodes. Further prospective studies with IGRT should be performed in patients with non-laryngeal and non-hypopharyngeal head and neck cancers to verify this hypothesis.
Sigma-Aldrich
Barium sulfate, ReagentPlus®, 99%
Sigma-Aldrich
Barium sulfate, 99.99% trace metals basis
Sigma-Aldrich
Barium sulfate, meets USP testing specifications