Merck
  • Catalytic desymmetrization of cyclohexadienes by asymmetric bromolactonization.
Catalytic desymmetrization of cyclohexadienes by asymmetric bromolactonization.
Organic letters (2012-11-15)
Kazutada Ikeuchi, Shunsuke Ido, Satoshi Yoshimura, Tomohiro Asakawa, Makoto Inai, Yoshitaka Hamashima, Toshiyuki Kan
ABSTRACT
Asymmetric bromolactonization of prochiral cyclohexadiene derivatives with N-bromosuccimide proceeded in the presence of (DHQD)(2)PHAL as a chiral catalyst to afford the corresponding bromolactones with up to 93% ee. This reaction was also applicable to the kinetic resolution of a racemic cyclic ene-carboxylic acid, where the starting material was recovered with high enantioselectivity.
Sigma-Aldrich
1,4-Cyclohexadiene, 97%
Sigma-Aldrich
1,4-Cyclohexadiene, purum, ≥97.0% (GC)