Merck
  • Simultaneous trace-levels determination of Hg(II) and Pb(II) ions in various samples using a modified carbon paste electrode based on multi-walled carbon nanotubes and a new synthesized Schiff base.
Simultaneous trace-levels determination of Hg(II) and Pb(II) ions in various samples using a modified carbon paste electrode based on multi-walled carbon nanotubes and a new synthesized Schiff base.
Analytica chimica acta (2012-09-15)
Abbas Afkhami, Hasan Bagheri, Hosein Khoshsafar, Mohammad Saber-Tehrani, Masoumeh Tabatabaee, Ali Shirzadmehr
ABSTRACT
A modified carbon paste electrode based on multi-walled carbon nanotubes (MWCNTs) and 3-(4-methoxybenzylideneamino)-2-thioxothiazolodin-4-one as a new synthesized Schiff base was constructed for the simultaneous determination of trace amounts of Hg(II) and Pb(II) by square wave anodic stripping voltammetry. The modified electrode showed an excellent selectivity and stability for Hg(II) and Pb(II) determinations and for accelerated electron transfer between the electrode and the analytes. The electrochemical properties and applications of the modified electrode were studied. Operational parameters such as pH, deposition potential and deposition time were optimized for the purpose of determination of traces of metal ions at pH 3.0. Under optimal conditions the limits of detection, based on three times the background noise, were 9.0×10(-4) and 6.0×10(-4) μmol L(-1) for Hg(II) and Pb(II) with a 90 s preconcentration, respectively. In addition, the modified electrode displayed a good reproducibility and selectivity, making it suitable for the simultaneous determination of Hg(II) and Pb(II) in real samples such as sea water, waste water, tobacco, marine and human teeth samples.
Sigma-Aldrich
Rhodanine, 97%
Supelco
p-Anisaldehyde, analytical standard
Sigma-Aldrich
p-Anisaldehyde, 98%
Sigma-Aldrich
p-Anisaldehyde, ≥97.5%, FCC, FG