Merck
  • Molecular characterization of colorectal cancer related peritoneal metastatic disease.
Molecular characterization of colorectal cancer related peritoneal metastatic disease.
Nature communications (2022-08-05)
Kristiaan J Lenos, Sander Bach, Leandro Ferreira Moreno, Sanne Ten Hoorn, Nina R Sluiter, Sanne Bootsma, Felipe A Vieira Braga, Lisanne E Nijman, Tom van den Bosch, Daniel M Miedema, Erik van Dijk, Bauke Ylstra, Ruth Kulicke, Fred P Davis, Nicolas Stransky, Gromoslaw A Smolen, Robert R J Coebergh van den Braak, Jan N M IJzermans, John W M Martens, Sally Hallam, Andrew D Beggs, Geert J P L Kops, Nico Lansu, Vivian P Bastiaenen, Charlotte E L Klaver, Maria C Lecca, Khalid El Makrini, Clara C Elbers, Mark P G Dings, Carel J M van Noesel, Onno Kranenburg, Jan Paul Medema, Jan Koster, Lianne Koens, Cornelis J A Punt, Pieter J Tanis, Ignace H de Hingh, Maarten F Bijlsma, Jurriaan B Tuynman, Louis Vermeulen
ABSTRACT
A significant proportion of colorectal cancer (CRC) patients develop peritoneal metastases (PM) in the course of their disease. PMs are associated with a poor quality of life, significant morbidity and dismal disease outcome. To improve care for this patient group, a better understanding of the molecular characteristics of CRC-PM is required. Here we present a comprehensive molecular characterization of a cohort of 52 patients. This reveals that CRC-PM represent a distinct CRC molecular subtype, CMS4, but can be further divided in three separate categories, each presenting with unique features. We uncover that the CMS4-associated structural protein Moesin plays a key role in peritoneal dissemination. Finally, we define specific evolutionary features of CRC-PM which indicate that polyclonal metastatic seeding underlies these lesions. Together our results suggest that CRC-PM should be perceived as a distinct disease entity.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-MSN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution