Merck
  • Inhibitory effect of UDP-glucose on cAMP generation and insulin secretion.
Inhibitory effect of UDP-glucose on cAMP generation and insulin secretion.
The Journal of biological chemistry (2020-08-29)
Fariborz Parandeh, Stefan Amisten, Gaurav Verma, Israa Mohammed Al-Amily, Pontus Dunér, Albert Salehi
ABSTRACT
Type-2 diabetes (T2D) is a global disease caused by the inability of pancreatic β-cells to secrete adequate insulin. However, the molecular mechanisms underlying the failure of β-cells to respond to glucose in T2D remains unknown. Here, we investigated the relative contribution of UDP-glucose (UDP-G), a P2Y14-specific agonist, in the regulation of insulin release using human isolated pancreatic islets and INS-1 cells. P2Y14 was expressed in both human and rodent pancreatic β-cells. Dose-dependent activation of P2Y14 by UDP-G suppressed glucose-stimulated insulin secretion (GSIS) and knockdown of P2Y14 abolished the UDP-G effect. 12-h pretreatment of human islets with pertussis-toxin (PTX) improved GSIS and prevented the inhibitory effect of UDP-G on GSIS. UDP-G on GSIS suppression was associated with suppression of cAMP in INS-1 cells. UDP-G decreased the reductive capacity of nondiabetic human islets cultured at 5 mm glucose for 72 h and exacerbated the negative effect of 20 mm glucose on the cell viability during culture period. T2D donor islets displayed a lower reductive capacity when cultured at 5 mm glucose for 72 h that was further decreased in the presence of 20 mm glucose and UDP-G. Presence of a nonmetabolizable cAMP analog during culture period counteracted the effect of glucose and UDP-G. Islet cultures at 20 mm glucose increased apoptosis, which was further amplified when UDP-G was present. UDP-G modulated glucose-induced proliferation of INS-1 cells. The data provide intriguing evidence for P2Y14 and UDP-G's role in the regulation of pancreatic β-cell function.
Sigma-Aldrich
Uridine 5′-diphosphoglucose disodium salt hydrate from Saccharomyces cerevisiae, ≥98%