Merck
  • Peptide-mediated cationic micelles drug-delivery system applied on a VEGFR3-overexpressed tumor.
Peptide-mediated cationic micelles drug-delivery system applied on a VEGFR3-overexpressed tumor.
Journal of materials chemistry. B (2019-02-21)
Qi Y Wang, Hong M Li, Zhi P Dong, Bing X Li, Ming Huo, Tao Lu, Yue Wang
ABSTRACT
Copolymers as a kind of drug delivery carrier always lack targeting efficiency. So a peptide conjugated to a drug delivery system has attracted much attention for tumor-targeted nanomedicine. Thus, we here report a conjugation compound consisting of a copolymer (PEG-b-PLL) and a peptide (Cys-Ile-Gln-Pro-Phe-Tyr-Pro, CP7). For receptor-mediated endocytosis by this peptide, the CP7-PEG-b-PLL conjugation significantly enhanced the chemotherapeutic efficacy as a potent nanocarrier compared with free DOX. The CP7-PEG-b-PLL exhibited excellent pharmacokinetic behavior via a radioactive iodine-131 (I) tracing method. With this, the CP7-PEG-b-PLL/DOX system showed better tumor growth inhibition when studied on A549 cell lines and subcutaneous tumor models, but with less toxicity than free DOX. All these results suggest that the CP7-modified drug cationic micelles could represent a novel platform for successful drug delivery toward VEGFR3-overexpressed tumors.
Sigma-Aldrich
Methoxy-poly(ethylene glycol) amine-block-poly-L-lysine hydrobromide, PEG average Mn 2000, PLLA average Mn 10000
Sigma-Aldrich
Methoxy-poly(ethylene glycol) amine-block-poly-L-lysine hydrobromide, PEG average Mn 2000, PLLA average Mn 5000