Merck
  • Chiroptical Spectroscopic Studies on Lanthanide Complexes with Valinamide Derivatives in Solution.
Chiroptical Spectroscopic Studies on Lanthanide Complexes with Valinamide Derivatives in Solution.
ChemPlusChem (2020-01-23)
Miki Hasegawa, Daichi Iwasawa, Takuma Kawaguchi, Hikaru Koike, Akira Saso, Shuhei Ogata, Ayumi Ishii, Hitomi Ohmagari, Munetaka Iwamura, Koichi Nozaki
ABSTRACT
Ligands based on 2,2'-bipyridine and valinamide moieties induce circularly polarized luminescence in their europium complexes. Both the R and S enantiomers of the complexes were successfully obtained. Single-crystal X-ray analysis of the racemic crystal confirmed that the ligand is coordinated to the europium ion in a tetradentate fashion. The π-electronic system of the ligand is co-planar with the valinamide moiety, and acts as an efficient photoantenna to sensitize europium luminescence by UV excitation. The luminescence quantum yield (QY) of europium in the valinamide-containing complex was 44 % in acetonitrile. The glum value to evaluate the circularly polarized luminescence was relatively high at |0.13| estimated from their magnetic dipole transitions around 593 nm. For comparison, we prepared hexadentate europium complexes in the S- and R-forms derived from two bipyridine moieties linked by ethylenediamines. The determined QYs were 18 % (S) and 16 % (R), and the glum value |0.12| for the hexadentate complexes. The photophysical properties of the gadolinium complexes of the ligands were also evaluated.
Sigma-Aldrich
L-Valinamide hydrochloride, 97%