Merck
  • Molecular localization techniques in the diagnosis and characterization of nonhuman primate infectious diseases.
Molecular localization techniques in the diagnosis and characterization of nonhuman primate infectious diseases.
Veterinary pathology (2013-10-31)
K G Mansfield, V G Sasseville, S V Westmoreland
ABSTRACT
Molecular localization techniques remain important diagnostic and research tools for the pathologist evaluating nonhuman primate tissues. In situ hybridization and immunohistochemistry protocols have been developed for many important pathogens of nonhuman primates, including RNA and DNA viruses, prions, and bacterial, protozoal, and fungal pathogens. Such techniques will remain critical in defining the impact and relevance of novel agents on animal health and disease. A comparative pathology perspective often provides valuable insight to the best strategy for reagent development and can also facilitate interpretation of molecular localization patterns. Such a perspective is grounded in a firm understanding of microbe-host pathobiology. This review summarizes current molecular localization protocols used in the diagnosis of selected primate infectious diseases.
Sigma-Aldrich
Anti-Measles Blend Antibody, hemagglutinin & matrix protein, clones CV1, CV4, CV8, CV9, ascites fluid, Chemicon®, from mouse
Sigma-Aldrich
Anti-Dengue Virus Complex Antibody, clone D3-2H2-9-21, clone D3-2H2-9-21, Chemicon®, from mouse
Sigma-Aldrich
Anti-Measles Blend Antibody, matrix protein, clones CV8, CV9, Chemicon®, from mouse