Merck
  • Dyes Adsorption Behavior of Fe3O4 Nanoparticles Functionalized Polyoxometalate Hybrid.
Dyes Adsorption Behavior of Fe3O4 Nanoparticles Functionalized Polyoxometalate Hybrid.
Molecules (Basel, Switzerland) (2019-08-31)
Jie Li, Chen Si, Haiyan Zhao, Qingxi Meng, Bowen Chang, Mingxue Li, Hongling Liu
ABSTRACT
The magnetic adsorbent, Fe3O4@[Ni(HL)2]2H2[P2Mo5O23]·2H2O (Fe3O4@1), is synthesized by employing the nanoparticles Fe3O4 and polyoxometalate hybrid 1. Zero-field-cooled (ZFC) and field-cooled (FC) curves show that the blocking temperature of Fe3O4@1 was at 120 K. Studies of Fe3O4@1 removing cationic and anionic dyes from water have been explored. The characterization of Fe3O4@1, effects of critical factors such as dosage, the concentration of methylene blue (MB), pH, adsorption kinetics, isotherm, the removal selectivity of substrate and the reusability of Fe3O4@1 were assessed. The magnetic adsorbent displayed an outstanding removal activity for the cationic dye at a broad range of pH. The adsorption kinetics and isotherm models revealed that the adsorption process of Fe3O4@1 was mainly governed via chemisorption. The maximum capacity of Fe3O4@1 adsorbing substance was 41.91 mg g-1. Furthermore, Fe3O4@1 showed its high stability by remaining for seven runs of the adsorption-desorption process with an effective MB removal rate, and could also be developed as a valuable adsorbent for dyes elimination from aqueous system.
Sigma-Aldrich
Dioctyl ether, 99%
Sigma-Aldrich
1,2-Hexadecanediol, technical grade, 90%
Sigma-Aldrich
2-Acetylpyrazine, ≥99%, FCC, FG