Merck
  • Mucin-1 is required for Coxsackie Virus B3-induced inflammation in pancreatitis.
Mucin-1 is required for Coxsackie Virus B3-induced inflammation in pancreatitis.
Scientific reports (2019-07-25)
Xiang Liu, Dahn L Clemens, James A Grunkemeyer, Jeffrey D Price, Kelly O'Connell, Nora M Chapman, Peter Storz, Haitao Wen, Jesse L Cox, Whitney L Reid, Michael A Hollingsworth, Sarah Thayer
ABSTRACT
The Muc-1 oncoprotein is a tumor-associated mucin often overexpressed in pancreatic cancer. We report that knockout of Muc-1 reduced the degree of pancreatic inflammation that resulted from infection with Coxsackievirus B3 (CVB3) in a mouse model. CVB3-infected Muc-1-deficient (Muc-1KO) mice had significantly reduced infiltration of macrophages into the murine pancreas. We found that Muc-1 signaling through NF-κB increased expression of ICAM-1, a pro-inflammatory mediator that recruits macrophages. Further investigation revealed that bone marrow derived macrophages (BMDM) from the Muc-1KO mice exhibited defective migration properties, in part due to low expression of the C-C motif chemokine receptor (CCR2) and the integrin Very Late Antigen 4 (VLA-4). The results presented here provide novel insight into the role of Muc-1 in regulating the inflammatory response and the cellular microenvironment in pancreatitis.
Sigma-Aldrich
Anti-Coxsackievirus B3 Antibody, clone 280-5F-4E-5E, clone 280-5F-4E-5E, Chemicon®, from mouse