Merck
  • The trifunctional protein mediates thyroid hormone receptor-dependent stimulation of mitochondria metabolism.
The trifunctional protein mediates thyroid hormone receptor-dependent stimulation of mitochondria metabolism.
Molecular endocrinology (Baltimore, Md.) (2012-05-10)
E Sandra Chocron, Naomi L Sayre, Deborah Holstein, Nuttawut Saelim, Jamal A Ibdah, Lily Q Dong, Xuguang Zhu, Sheue-Yann Cheng, James D Lechleiter
ABSTRACT
We previously demonstrated that the thyroid hormone, T(3), acutely stimulates mitochondrial metabolism in a thyroid hormone receptor (TR)-dependent manner. T(3) has also recently been shown to stimulate mitochondrial fatty acid oxidation (FAO). Here we report that TR-dependent stimulation of metabolism is mediated by the mitochondrial trifunctional protein (MTP), the enzyme responsible for long-chain FAO. Stimulation of FAO was significant in cells that expressed a nonnuclear amino terminus shortened TR isoform (sTR(43)) but not in adult fibroblasts cultured from mice deficient in both TRα and TRβ isoforms (TRα(-/-)β(-/-)). Mouse embryonic fibroblasts deficient in MTP (MTP(-/-)) did not support T(3)-stimulated FAO. Inhibition of fatty-acid trafficking into mitochondria using the AMP-activated protein kinase inhibitor 6-[4-(2-piperidin-1-yl-ethoxy)-phenyl)]-3-pyridin-4-yl-pyrrazolo[1,5-a]-pyrimidine (compound C) or the carnitine palmitoyltransferase 1 inhibitor etomoxir prevented T(3)-stimulated FAO. However, T(3) treatment could increase FAO when AMP-activated protein kinase was maximally activated, indicating an alternate mechanism of T(3)-stimulated FAO exists, even when trafficking is presumably high. MTPα protein levels and higher molecular weight complexes of MTP subunits were increased by T(3) treatment. We suggest that T(3)-induced increases in mitochondrial metabolism are at least in part mediated by a T(3)-shortened TR isoform-dependent stabilization of the MTP complex, which appears to lower MTP subunit turnover.
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
AICA-Riboside, AICA-Riboside, CAS 2627-69-2, is a cell-permeable nucleoside compound whose phosphorylated metabolite activates AMPK and acts as a regulator of de novo purine synthesis.
Sigma-Aldrich
n-Dodecyl β-D-maltoside, ≥98% (GC)
Sigma-Aldrich
Albumin, Bovine Serum, Fraction V, Fatty Acid-Free
Sigma-Aldrich
AMPK Inhibitor, Compound C, InSolution, ≥95%, 10 mM, AMPK Inhibitor