Merck
  • PMN-MDSC and arginase are increased in myeloma and may contribute to resistance to therapy.
PMN-MDSC and arginase are increased in myeloma and may contribute to resistance to therapy.
Expert review of molecular diagnostics (2018-05-01)
Alessandra Romano, Nunziatina Laura Parrinello, Piera La Cava, Daniele Tibullo, Cesarina Giallongo, Giuseppina Camiolo, Fabrizio Puglisi, Marina Parisi, Maria Cristina Pirosa, Enrica Martino, Concetta Conticello, Giuseppe Alberto Palumbo, Francesco Di Raimondo
ABSTRACT
Despite improvement in overall response due to the introduction of the first-in-class proteasome inhibitor bortezomib (btz), multiple myeloma (MM) is still an incurable disease due to the immune-suppressive bone marrow (BM) environment. Thus, the authors aimed to identify the role of CD11b+CD15+CD14-HLA-DR- granulocytic-like myeloid-derived suppressor cells (PMN-MDSC) in MM patients treated up-front with novel agents. In MM cell lines and primary cells derived by patients affected by MGUS and MM, we investigated sensitivity to bortezomib and lenalidomide in presence of Arg-1 and PMN-MDSC. The authors found that PMN-MDSC and their function through increased arginase-1 (Arg-1) are associated with MM progression. When the authors assessed cell viability of the human myeloma cell lines MM1.s, OPM2 and U266 treated with 5-20 nM btz for 24 h in PMN-MDSC conditioned media, they disclosed that amount of Arg-1 and Arg-1 inhibition could affect btz sensitivity in-vitro. PMN-MDSC and Arg-1 were increased in peripheral blood of newly diagnosed MM patients compared to healthy subjects. PMN-MDSC and arginase were reduced after exposure to lenalidomide-based regimen but increased after btz-based treatment. In MM, Arg-1 is mainly expressed by PMN-MDSC. PMN-MDSC and Arg-1 are reduced in vivo after lenalidomide but not bortezomib treatment.
Sigma-Aldrich
BEC hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%