
Tiliroside

Synonym(s):
Astragalin 6"-trans-p-coumarate, Kaempferol 3-O-β-D-(6″-O-(E)-p-coumaroyl)glucopyranoside, Potengriffioside A
Empirical Formula (Hill Notation):
C30H26O13
CAS Number:
Molecular Weight:
594.52
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing