
Pyrex® low actinic media storage bottle, graduated with screw cap

Synonym(s):
Corning® 51395
Products
Product Category
Manufacturer
Capacity (mL)
Diameter (mm)
Height (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Sterility
Showing 1-6 of 6
Compare
Product Number
Description
Pricing