
Proguanil Related Compound A

Synonym(s):
1-Cyano-3-isopropylguanidine, N-Cyano-N′-(1-methylethyl)guanidine, NSC 643485
Empirical Formula (Hill Notation):
C5H10N4
CAS Number:
Molecular Weight:
126.16
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2