
Pinoxaden Metabolite SYN502836

Synonym(s):
3,5-Diethyl-4-(hexahydro-7,9-dioxo-7H-pyrazolo[1,2-d][1,4,5]oxadiazepin-8-yl)-benzoic acid
Empirical Formula (Hill Notation):
C18H22N2O5
CAS Number:
Molecular Weight:
346.38
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing