
Physostigmine salicylate

Synonym(s):
Eserine salicylate salt, Physostigmine salicylate salt
Empirical Formula (Hill Notation):
C15H21N3O2 · C7H6O3
CAS Number:
Molecular Weight:
413.47
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-2 of 2