
Parteck® LUB STA 50

Synonym(s):
Palmitic acid – stearic acid mixture, Fatty acids C16-18, Stearic acid - palmitic acid mixture
CAS Number:
Products
Product Category
Brand
Physical Form
Product Line
Agency
Quality Segment
Application
Particle Size (µm)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing