
o-Phosphoric Acid

Synonym(s):
Phosphoric acid, Orthophosphoric acid
Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Agency
Quality Segment
Markush Class
Markush Group
Showing 1-3 of 3