
Nordazepam

Synonym(s):
7-Chloro-1,3-dihydro-5-phenyl-2H-1,4-benzodiazepin-2-one, Desmethyldiazepam
Empirical Formula (Hill Notation):
C15H11ClN2O
CAS Number:
Molecular Weight:
270.71
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing