
Montelukast sodium

Synonym(s):
Montelukast sodium
Empirical Formula (Hill Notation):
C35H35ClNO3S·Na
CAS Number:
Molecular Weight:
608.17
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique