
ML348

Synonym(s):
N-[2-Chloro-5-(trifluoromethyl)phenyl]-2-[4-(2-furoyl)-1-piperazinyl]acetamide
Empirical Formula (Hill Notation):
C18H17ClF3N3O3
CAS Number:
Molecular Weight:
415.79
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing