Skip to Content
Merck
Methyl propionate analytical standard

Methyl propionate

Methyl propionate analytical standard
Linear Formula:
CH3CH2COOCH3
CAS Number:
Molecular Weight:
88.11
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing