
Methyl cedryl ketone

Empirical Formula (Hill Notation):
C17H26O
CAS Number:
Molecular Weight:
246.39
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing