
Linezolid R-Isomer

Synonym(s):
N-[[(5R)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]acetamide, (R)-Linezolid
Empirical Formula (Hill Notation):
C16H20FN3O4
CAS Number:
Molecular Weight:
337.35
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2