
Hesperetin

Synonym(s):
3′,5,7-Trihydroxy-4′-methoxyflavanone, 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4-chromanone
Empirical Formula (Hill Notation):
C16H14O6
CAS Number:
Molecular Weight:
302.28
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing