Edrophonium chloride
Synonym(s):
Dimethylethyl(3-hydroxyphenyl)ammonium chloride, Edrophone chloride, Ethyl(m-hydroxyphenyl)dimethylammonium chloride, N-Ethyl-3-hydroxy-N,N-dimethyl-benzenaminium chloride
Empirical Formula (Hill Notation):
C10H16ClNO
CAS Number:
Molecular Weight:
201.69
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-3 of 3