
Duloxetine Related Compound H

Synonym(s):
(S)-4-{Methyl[3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propyl]amino}-4-oxobutanoic acid
Empirical Formula (Hill Notation):
C22H23NO4S
CAS Number:
Molecular Weight:
397.49
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2