![Dimethomorph Mixture of E + Z isomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/377/559/626713cd-c2c8-46ea-bf1c-96d39a5509dd/640/626713cd-c2c8-46ea-bf1c-96d39a5509dd.png)
Dimethomorph
![Dimethomorph Mixture of E + Z isomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/377/559/626713cd-c2c8-46ea-bf1c-96d39a5509dd/640/626713cd-c2c8-46ea-bf1c-96d39a5509dd.png)
Empirical Formula (Hill Notation):
C21H22ClNO4
CAS Number:
Molecular Weight:
387.86
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2