Skip to Content
Merck
Deblock Reagent 10% DCA in Toluene Biosynthesis Novabiochem®

Deblock Reagent 10% DCA in Toluene

Deblock Reagent 10% DCA in Toluene Biosynthesis Novabiochem®
Synonym(s):
DCA Deblock
Empirical Formula (Hill Notation):
C2H2Cl2O2
CAS Number:
Molecular Weight:
128.94

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing