
Aminoethoxyvinyl glycine hydrochloride

Synonym(s):
(S)-trans-2-Amino-4-(2-aminoethoxy)-3-butenoic acid hydrochloride, L-α-(2-Aminoethoxyvinyl)glycine hydrochloride, AVG-HCl
Empirical Formula (Hill Notation):
C6H12N2O3 · HCl
CAS Number:
Molecular Weight:
196.63
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing