
4-Chloro-α-cyanocinnamic acid

Synonym(s):
3-(4-Chlorophenyl)-2-cyano-2-propenoic acid, 3-(4-Chlorophenyl)-2-cyanoacrylic acid, p-Chloro-α-cyanocinnamic acid, p-Chlorobenzylidenecyanoacetic acid, ClCCA, NSC 2479
Empirical Formula (Hill Notation):
C10H6ClNO2
CAS Number:
Molecular Weight:
207.61
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing