Skip to Content
MO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-(4-carboxy-phenyl)-piperazine-1-carboxylic acid
tert
-butyl ester
Empirical Formula (Hill Notation):
C
16
H
22
N
2
O
4
CAS Number:
162046-66-4
Molecular Weight:
306.36
Products
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
ANV00053
4-(4-carboxy-phenyl)-piperazine-1-carboxylic acid
tert
-butyl ester,
Aldrich
CPR
Expand
View Pricing
CDS007031
4-(4-carboxy-phenyl)-piperazine-1-carboxylic acid
tert
-butyl ester,
Aldrich
CPR
Expand
View Pricing