Merck
Photocatalysts
Photoreactor in organic synthesis lab for photoredox catalysis
Photoredox catalysis is a powerful tool in organic chemical synthesis that utilizes visible light to power a chemical reaction. Widespread adoption of visible light photoredox catalysis has been dependent on access to photocatalysts and a reliable light source. We’re pleased to support your photocatalysis endeavors to create new bonds and rapidly assemble complex products.  
Chemists have long struggled with reproducibility in photoredox chemistry. Both varied reaction setups and individual reactions performed with the same setup can be tricky. Our new labware seeks to alleviate these issues by providing photoreactors for each stage of reaction development, while ensuring high levels of consistency across photocatalytic reactions and between runs.
Products
Sort by Relevance
Showing 1-20 of 78
Change View
Showing 1-20 of 78
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 78 results
 Our photoreactors serve as reliable visible light sources  for a variety of applications and reaction scales:  
  • The Photo KitAlysis starter kit is perfect for screening of 24 micro-scale simultaneous photocatalytic reactions for reaction optimization.
  • The SynLED Parallel Photoreactor enables small-scale reactions for rapid library generation by running 16 simultaneous reactions in 2 dram vials.
  • Penn PhD combines LED illumination, mechanical stirring, and cooling into one device while accepting reaction vials from 1 mL up to 40 mL, allowing chemists to streamline synthetic sequences.
  • The Bio-Photoreactor BPR200 utilizes LED illumination and a self-cooling heat sink for applications in parallel photoredox reactions, covalent labeling of biomolecules, and experiments using living cells.
  • The LightOx PhotoReact 365 provides UV illumination suitable for photochemical and photobiological high-throughput screening reactions.
Please refer to our photocatalysis technical article to determine which photoreactor is most suitable for your needs.
With a continuously growing portfolio of acridinium, iridium and ruthenium catalysts, and other metal-free organic photocatalysts and ligands, we provide a broad range of complex photoredox catalysts for any reaction design. Our KitAlysis™ screening kits enable chemists to quickly and efficiently find good reaction conditions for a wide range of photoredox catalyzed reactions.
Related Product Resources
  • Visible-light Photoredox Catalysis
    The application of radical chemistry towards organic synthesis is well-developed and wide-reaching, though often hampered by a dependence on toxic radical initiators.
  • Photoredox Catalysis Guide
    Sign up for your complimentary Photoredox Catalysis Guide featuring key examples of previously inaccessible bond formations, standard reaction setups, and catalysts references and properties.
  • Illuminating Research with the LightOx PhotoReact 365 Benchtop Photoreactor
    Photochemical and photobiological research is a rapidly growing field with unique experimental requirements, and achieving a reliable and reproducible means of sample irradiation is often just as much a challenge as performing the research itself.