SML2568
XMD8-87
≥98% (HPLC)
Synonym(s):
5,11-Dihydro-2-[[2-methoxy-4-(4-methyl-1-piperazinyl)phenyl]amino]-11-methyl-6H-pyrimido[4,5-b][1,4]benzodiazepin-6-one, ACK1-B19
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C24H27N7O2
CAS Number:
Molecular Weight:
445.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
XMD8-87 is a potent and selective inhibitor of Ack1 (Activated CDC42 kinase 1, TNK2), a kinase that regulates cellular attachment and migration and whose over-expression correlates with a more invasive phenotype of cancer cells and predisposition of primary tumor cells to metastasis. XMD8-87 had a Kd of 15 nM against ACK1 and was also found to inhibit murine Ba/F3 tumor cells having leukemia-associated TNK2 mutations. It inhibited TNK2 D163E cells with an IC50 value of 38 nM and TNK2 R806Q cells with an IC50 value of 113 nM.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chandrasekhar V Miduturu et al.
Chemistry & biology, 18(7), 868-879 (2011-08-02)
Selective protein kinase inhibitors have only been developed against a small number of kinase targets. Here we demonstrate that "high-throughput kinase profiling" is an efficient method for the discovery of lead compounds for established as well as unexplored kinase targets.
Development and Repurposing of Small-Molecule Kinase Inhibitors to Target Novel Leukemogenic TNK2 Mutations
Maxson JE., Melissa AL, Wang J, Deng X, Luty SB, Sun H, Gorenstein J, Middleton R, Gray NS, et al
Blood, 124, 435-435 (2014)
Julia E Maxson et al.
Cancer research, 76(1), 127-138 (2015-12-19)
The amount of genomic information about leukemia cells currently far exceeds our overall understanding of the precise genetic events that ultimately drive disease development and progression. Effective implementation of personalized medicine will require tools to distinguish actionable genetic alterations within
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service