Skip to Content
Merck
All Photos(1)

Key Documents

SCP0165

Sigma-Aldrich

Hepatitis C Virus NS3 Protease Substrate, fluorogenic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H53N7O13S1
Molecular Weight:
847.93
UNSPSC Code:
12352202
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥73%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Glu-Glu-Val-Val-Ala-Cys-AMC

Application

Hepatitis C virus (HCV) NS3 protease is the key enzyme for virus maturation. AcEEVVAC-AMC is used as a fluorogenic substrate of NS3 protease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service