GC Analysis of Background Levels of a Carbopack™ X Thermal Desorption Tube on SPB®-1
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SPB-1, 60 m x 0.25 mm I.D., 1.0 μm (24031)
oven
35 °C (8 min), 5 °C/min to 100 °C, 15 °C/min to 230 °C (hold 4 min)
inj. temp.
200 °C
MSD interface
230 °C
scan range
m/z = 35-260
carrier gas
nitrogen, 2 mL/min. constant
injection
split 10:1
Description
Legal Information
Carbopack is a trademark of Merck KGaA, Darmstadt, Germany
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany