Merck
  • Generation of Lactobacillus plantarum strains with improved potential to target gastrointestinal disorders related to sugar malabsorption.
Generation of Lactobacillus plantarum strains with improved potential to target gastrointestinal disorders related to sugar malabsorption.
Food research international (Ottawa, Ont.) (2017-03-16)
Helena Šeme, Bojana Bogovič Matijašić, Karmen Švigelj, Tomaž Langerholc, Štefan Fujs, Jaka Horvat, Emil Zlatić, Krešimir Gjuračić, Hrvoje Petković, Mateja Štempelj, Blaženka Kos, Jagoda Šušković, Gregor Kosec
ABSTRACT
Malabsorption of dietary sugars is a common cause of gastrointestinal discomfort, affecting up to one in three people with debilitating symptoms, such as abdominal pain, osmotic diarrhoea, bloating and flatulence. Besides dietary interventions, it has been suggested that ingestion of lactobacilli may alleviate these symptoms. The objectives of this study were to generate strains with improved potential to ameliorate sugar malabsorption related gastrointestinal disorders. Initial selection was made from 183 natural isolates of lactic acid bacteria, on the basis of broad sugar fermentation ability, absence of gas production, gastrointestinal survival and susceptibility to important medical antimicrobials. Two strains of L. plantarum (KR6 and M5) exhibited favourable characteristics for all criteria, and were further optimised through random mutagenesis and selection approaches. Ultraviolet light (UV) exposure resulted in mutants characterized by better survival (for 1.9 log and 1.4 log) in gastrointestinal conditions. Subsequent exposure to ethyl methanesulfonate (EMS) provided mutants with greater tolerance to glucose induced catabolic repression. UV and UV-EMS mutants of L. plantarum M5 showed improved adhesion ability. As a result of this optimisation, L. plantarum MP2026 and L. plantarum MP2420 have been identified as promising candidates for probiotics, intended for alleviation of gastrointestinal discomfort originating from sugar malabsorption.
Product Description
Millipore
Bile bovine, dried, unfractionated