Merck
  • An alternative mode of CD43 signal transduction activates pro-survival pathways of T lymphocytes.
An alternative mode of CD43 signal transduction activates pro-survival pathways of T lymphocytes.
Immunology (2016-09-09)
Maria Elena Bravo-Adame, Rosario Vera-Estrella, Bronwyn J Barkla, Cecilia Martínez-Campos, Angel Flores-Alcantar, Jose Pablo Ocelotl-Oviedo, Gustavo Pedraza-Alva, Yvonne Rosenstein
ABSTRACT
CD43 is one of the most abundant co-stimulatory molecules on a T-cell surface; it transduces activation signals through its cytoplasmic domain, contributing to modulation of the outcome of T-cell responses. The aim of this study was to uncover new signalling pathways regulated by this sialomucin. Analysis of changes in protein abundance allowed us to identify pyruvate kinase isozyme M2 (PKM2), an enzyme of the glycolytic pathway, as an element potentially participating in the signalling cascade resulting from the engagement of CD43 and the T-cell receptor (TCR). We found that the glycolytic activity of this enzyme was not significantly increased in response to TCR+CD43 co-stimulation, but that PKM2 was tyrosine phosphorylated, suggesting that it was performing moonlight functions. We report that phosphorylation of both Y105 of PKM2 and of Y705 of signal transducer and activator of transcription 3 was induced in response to TCR+CD43 co-stimulation, resulting in activation of the mitogen-activated protein kinase kinase 5/extracellular signal-regulated kinase 5 (MEK5/ERK5) pathway. ERK5 and the cAMP response element binding protein (CREB) were activated, and c-Myc and nuclear factor-κB (p65) nuclear localization, as well as Bad phosphorylation, were augmented. Consistent with this, expression of human CD43 in a murine T-cell hybridoma favoured cell survival. Altogether, our data highlight novel signalling pathways for the CD43 molecule in T lymphocytes, and underscore a role for CD43 in promoting cell survival through non-glycolytic functions of metabolic enzymes.
Sigma-Aldrich
XMD8-92, ≥98% (HPLC)
Sigma-Aldrich
PP2, ≥98% (HPLC)