Merck
  • Is there a role for rifampicin, ofloxacin and minocycline (ROM) therapy in the treatment of leprosy? Systematic review and meta-analysis.
Is there a role for rifampicin, ofloxacin and minocycline (ROM) therapy in the treatment of leprosy? Systematic review and meta-analysis.
Tropical medicine & international health : TM & IH (2011-09-15)
Maninder S Setia, Santosh S Shinde, Hemangi R Jerajani, Jean-François Boivin
ABSTRACT
A combination of rifampicin, ofloxacin and minocycline (ROM) is one of the newer recommendations for treatment of leprosy. We performed a systematic review and a meta-analysis of studies that had evaluated the efficacy of ROM therapy in treatment of paucibacillary and multibacillary leprosy patients. Studies were identified by searching the PubMed, Embase, LILACS and Cochrane databases. Data were abstracted from all relevant studies, and fixed effects models were used to calculate the summary estimate of effect in paucibacillary and multibacillary leprosy patients. Six studies comparing ROM therapy to multidrug therapy and eight studies that evaluated the effect of ROM therapy alone (no comparison group) were included in the review and meta-analysis. The combined estimate for single dose ROM vs. multidrug therapy in paucibacillary leprosy patients suggested that ROM was less effective than multidrug therapy in these patients [relative risk: 0.91, 95% confidence intervals (CI): 0.86-0.97]. However, the combined estimate for multiple doses of ROM vs. multidrug therapy in multibacillary leprosy patients suggested that ROM was as effective as multidrug therapy in reducing bacillary indices in these patients (proportion change: -4%, 95% CI -31% to 23%). No major side effects were reported in either the ROM or the multidrug treatment groups. Single-dose ROM therapy was less effective than multidrug therapy in paucibacillary patients. However, there are insufficient data to come to a valid conclusion on the efficacy of multidose ROM therapy in multibacillary leprosy, and additional studies with ROM therapy in multibacillary leprosy are needed. Furthermore, multiple doses may be considered as another alternative even for paucibacillary patients, and randomised controlled trials of this therapy may be useful to understand its contribution in the treatment and control of leprosy.
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Ofloxacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ofloxacin, VETRANAL®, analytical standard
Rifampicin, European Pharmacopoeia (EP) Reference Standard
USP
Ofloxacin, United States Pharmacopeia (USP) Reference Standard