Merck
  • Antifungal activity of strawberry fruit volatile compounds against Colletotrichum acutatum.
Antifungal activity of strawberry fruit volatile compounds against Colletotrichum acutatum.
Journal of agricultural and food chemistry (2007-06-15)
Francisco T Arroyo, Javier Moreno, Paula Daza, Lidiya Boianova, Fernando Romero
ABSTRACT
Eight volatile products characterizing strawberry aroma, which is generated from the oxidative degradation of linoleic and linolenic acids by a lipoxygenase (LOX) pathway, were examined because of their antifungal activity against Colletotrichum acutatum, one of the causal agents of strawberry anthracnose. In this study, the effects of aldehydes, alcohols, and esters on mycelial growth and conidia development were evaluated. (E)-Hex-2-enal was found to be the best inhibitor of mycelial growth [MID (minimum inhibitory doses)=33.65 microL L(-1)] and of spore germination (MID=6.76 microL L(-1)), while hexyl acetate was the least effective of all volatile compounds tested (MID=6441.89 microL L(-1) for mycelial growth and MID=1351.35 microL L(-1) for spore germination). Furthermore, the antifungal activity of (E)-hex-2-enal on susceptibility of strawberry fruits to C. acutatum was also confirmed. The presence of these molecules in jars containing strawberry fruits inoculated with a suspension of spores inhibited the fungus growth and prevented the appearance of symptoms. Moreover, a study of the effects of (E)-hex-2-enal on conidial cells of C. acutatum was evaluated by transmission electron microscopy. This volatile compound altered the structures of the cell wall and plasma membrane, causing disorganization and lysis of organelles and, eventually, cell death.
Sigma-Aldrich
Hexyl acetate, natural, ≥98%, FCC, FG
Supelco
Hexobarbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®