Merck
  • Serotonin neurotoxicity in rats after combined treatment with a dopaminergic agent followed by a nonneurotoxic 3,4-methylenedioxymethamphetamine (MDMA) analogue.
Serotonin neurotoxicity in rats after combined treatment with a dopaminergic agent followed by a nonneurotoxic 3,4-methylenedioxymethamphetamine (MDMA) analogue.
Pharmacology, biochemistry, and behavior (1991-12-01)
M P Johnson, X M Huang, D E Nichols
ABSTRACT
There is increasing evidence linking dopamine (DA) to the long-term serotonergic (5-HT) neurotoxic effects of certain substituted amphetamines such as 3,4-methylenedioxymethamphetamine (MDMA). The present study was undertaken to examine the importance of DA metabolism, uptake inhibition and release in the long-term effects of these drugs by combining various dopaminergic agents with an analogue of MDMA that had low neurotoxic liability, namely 5,6-methylenedioxy-2-aminoindan (MDAI). Monoamine and metabolite levels and the number of 5-HT uptake sites (using [3H]paroxetine binding) were determined 3 hours or 1 week after treatments. Combining the monoamine oxidase inhibitors, clorgyline (MAOA selective) or deprenyl (MAOB selective) with MDAI did not result in any long-term reductions of serotonergic markers. Similarly, combining the DA uptake inhibitor GBR-12909 with MDAI did not result in any long-term changes in monoamine levels at 1 week. In contrast, a single pretreatment of posttreatment with the nonvesicular DA releaser S-amphetamine and MDAI resulted in small but significant long-term changes in monoamine levels. More importantly, if a subacute dosing regimen (every 12 hours for 4 days) was utilized, the combination of S-amphetamine with MDAI resulted in a marked long-term decrease in the levels of cortical, hippocampal and striatal 5-HT, 5-HIAA and the number of 5-HT uptake sites. The results are discussed in terms of the significance of DA and especially nonvesicular DA release in the long-term effects of MDMA-like drugs.
Product Description
Supelco
MDAI hydrochloride solution, 100 μg/mL in acetonitrile: water (9:1) with 5% 1 M HCl (as free base), ampule of 1 mL, certified reference material, Cerilliant®