Merck
  • 12S-Lipoxygenase is necessary for human vascular smooth muscle cell survival.
12S-Lipoxygenase is necessary for human vascular smooth muscle cell survival.
Experimental cell research (2013-04-13)
G Weisinger, M Grafi-Cohen, M Hirsh, E Knoll, O Sharon, A Many, R Limor, N Stern
ABSTRACT
Considerable evidence has been published demonstrating the importance of lipoxygenase enzymes for vascular smooth muscle cell (VSMC) growth. The current study sets out to determine whether or not 12-lipoxygenase (12LO) is also important for human placental VSMC survival. Both a pharmacological and two 12LO antisense knockdown approaches were applied. The 12LO inhibitor baicalien induced a 2-2.5-fold increase in cell death, which appeared to result from apoptosis, as indicated by DNA fragmentation, activation of procaspase 3 to caspase 3 and cytochrome C release from the mitochondria to the cytosol. This apoptosis could be prevented by treatment with the 12LO product, 12 hydroxyeicosatetraenoic acid (12HETE). Human platelet-type 12LO-antisense knockdown, by either plasmid transfection or adeno-associated virus (AAV) infection also induced substantial VSMC death over controls, which could also be prevented by treatment with 12HETE, but not 5HETE. Hence, biochemical 12LO inhibition or 12LO-antisense knockdown in VSMC can induce programmed cell death. These observations suggest a previously unrecognized association between human VSMC survivability and 12LO.
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c from equine heart, suitable for GFC marker, BioReagent
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Cytochrome c from pigeon breast muscle, ≥95% based on Mol. Wt. 12,173 basis
Sigma-Aldrich
Baicalein, 98%
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis
Sigma-Aldrich
ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation