Merck
  • Lupeol supplementation improves blood pressure and lipid metabolism parameters in stroke-prone spontaneously hypertensive rats.
Lupeol supplementation improves blood pressure and lipid metabolism parameters in stroke-prone spontaneously hypertensive rats.
Bioscience, biotechnology, and biochemistry (2012-01-11)
Ardiansyah, Eri Yamaguchi, Hitoshi Shirakawa, Keishi Hata, Kazuyuki Hiwatashi, Kousaku Ohinata, Tomoko Goto, Michio Komai
ABSTRACT
Supplementation with lupeol (0.67 g·kg(-1)) of the AIN-93M-based diet fed for 7 weeks to stroke-prone spontaneously hypertensive rats caused significantly decreased blood pressure as compared with a control group. Urinary 8-hydroxy-2'-deoxyguanosine was significantly lower in the lupeol group. Finally, lupeol suppressed the hepatic mRNA expression levels of the genes involved in triglyceride and cholesterol synthesis.
Product Description
Supelco
Lupeol, analytical standard
Sigma-Aldrich
Lupeol, ≥94%