Merck
  • The effect of isothiocyanates on CYP1A1 and CYP1A2 activities induced by polycyclic aromatic hydrocarbons in Mcf7 cells.
The effect of isothiocyanates on CYP1A1 and CYP1A2 activities induced by polycyclic aromatic hydrocarbons in Mcf7 cells.
Toxicology in vitro : an international journal published in association with BIBRA (2009-04-14)
Katarzyna Skupinska, Irena Misiewicz-Krzeminska, Katarzyna Lubelska, Teresa Kasprzycka-Guttman
ABSTRACT
Polycyclic aromatic hydrocarbons (PAHs)--environmental carcinogens--are metabolized by CYP1A1 and CYP1A2 enzymes to oxy-derivatives, which are able to bind to DNA and initiate carcinogenesis. PAHs induce CYP1A1 and CYP1A2 activity, which increases the risk of development of carcinogenesis. Isothiocyanates (ITCs), naturally occurring in Brassica vegetables, possess chemopreventive properties and are able to reduce the CYP1A enzyme activity. In this paper we report our study of the ability of ITCs: sulforaphane and its analogues: isothiocyanate-2-oxohexyl and alyssin, to inhibit CYP1A1 and CYP1A2 enzyme activity induced by the PAHs, anthracene (ANT) and dibenzo[a,h]anthracene (DBA) in human breast cancer cell line Mcf7. The aim was to determine whether the differences in structure of ITCs change their inhibitory properties, and whether these properties depend on the type of inducer. The results indicate that the properties of ITCs depend on the type of PAH: ITCs are more potent in inhibiting activity induced by the weaker inducer. It was also found that the change in ITCs' structure influences their activities. ITC 2-oxohexyl was the weakest inhibitor, whereas sulforaphane and alyssin exhibited similar potency. The study revealed that inhibition of CYP1A1 activity is direct whereas inhibition of CYP1A2 activity is not only direct but is also caused by the level of protein disturbance.
Product Description
Dibenz[a,h]anthracene, BCR®, certified reference material
Supelco
Dibenz[a,h]anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland