Merck
  • Microglia and astrocytes mediate synapse engulfment in a MER tyrosine kinase-dependent manner after traumatic brain injury.
Microglia and astrocytes mediate synapse engulfment in a MER tyrosine kinase-dependent manner after traumatic brain injury.
Neural regeneration research (2023-02-09)
Hui Shen, Xiao-Jing Shi, Lin Qi, Cheng Wang, Muyassar Mamtilahun, Zhi-Jun Zhang, Won-Suk Chung, Guo-Yuan Yang, Yao-Hui Tang
ABSTRACT
Recent studies have shown that microglia/macrophages and astrocytes can mediate synaptic phagocytosis through the MER proto-oncokinase in developmental or stroke models, but it is unclear whether the same mechanism is also active in traumatic brain injury. In this study, we established a mouse model of traumatic brain injury and found that both microglia/macrophages and astrocytes phagocytosed synapses and expression of the MER proto-oncokinase increased 14 days after injury. Specific knockout of MER in microglia/macrophages or astrocytes markedly reduced injury volume and greatly improved neurobehavioral function. In addition, in both microglia/macrophages-specific and astrocytes-specific MER knock-out mice, the number of microglia/macrophage and astrocyte phagocytosing synapses was markedly decreased, and the total number of dendritic spines was increased. Our study suggested that MER proto-oncokinase expression in microglia/macrophages and astrocytes may play an important role in synaptic phagocytosis, and inhibiting this process could be a new strategy for treating traumatic brain injury.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Anti-LAMP-2 Antibody, clone ABL-93, clone ABL-93, from rat