Merck
  • CD36 mediates SARS-CoV-2-envelope-protein-induced platelet activation and thrombosis.
CD36 mediates SARS-CoV-2-envelope-protein-induced platelet activation and thrombosis.
Nature communications (2023-08-22)
Zihan Tang, Yanyan Xu, Yun Tan, Hui Shi, Peipei Jin, Yunqi Li, Jialin Teng, Honglei Liu, Haoyu Pan, Qiongyi Hu, Xiaobing Cheng, Junna Ye, Yutong Su, Yue Sun, Jianfen Meng, Zhuochao Zhou, Huihui Chi, Xuefeng Wang, Junling Liu, Yong Lu, Feng Liu, Jing Dai, Chengde Yang, Saijuan Chen, Tingting Liu
ABSTRACT
Aberrant coagulation and thrombosis are associated with severe COVID-19 post-SARS-CoV-2 infection, yet the underlying mechanism remains obscure. Here we show that serum levels of SARS-CoV-2 envelope (E) protein are associated with coagulation disorders of COVID-19 patients, and intravenous administration of the E protein is able to potentiate thrombosis in mice. Through protein pull-down and mass spectrometry, we find that CD36, a transmembrane glycoprotein, directly binds with E protein and mediates hyperactivation of human and mouse platelets through the p38 MAPK-NF-κB signaling pathway. Conversely, the pharmacological blockade of CD36 or p38 notably attenuates human platelet activation induced by the E protein. Similarly, the genetic deficiency of CD36, as well as the pharmacological inhibition of p38 in mice, significantly diminishes E protein-induced platelet activation and thrombotic events. Together, our study reveals a critical role for the CD36-p38 axis in E protein-induced platelet hyperactivity, which could serve as an actionable target for developing therapies against aberrant thrombotic events related to the severity and mortality of COVID-19.
Sigma-Aldrich
Goat Anti-Mouse IgM µ chain Antibody, Cy5 conjugate, Species Adsorbed, 1 mg/mL, Chemicon®
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Cy3 conjugate, Chemicon®, from goat