Merck
  • Drought-Induced Xylem Embolism Limits the Recovery of Leaf Gas Exchange in Scots Pine.
Drought-Induced Xylem Embolism Limits the Recovery of Leaf Gas Exchange in Scots Pine.
Plant physiology (2020-08-21)
Romy Rehschuh, Angelica Cecilia, Marcus Zuber, Tomáš Faragó, Tilo Baumbach, Henrik Hartmann, Steven Jansen, Stefan Mayr, Nadine Ruehr
ABSTRACT
Climate change increases the occurrence of prolonged drought periods with large implications for forest functioning. Scots pine (Pinus sylvestris) is one of the most abundant conifers worldwide, and evidence is rising that its resilience to severe drought is limited. However, we know little about its ability to recover from drought-induced embolism. To analyze postdrought hydraulic recovery, we investigated stress and recovery dynamics of leaf gas exchange, nonstructural carbohydrates, and hydraulic properties in 2.5-year-old Scots pine seedlings. We quantified the degree of xylem embolism by combining in vivo x-ray microtomography with intrusive techniques including measurements of hydraulic conductivity and dye staining during drought progression and short-term (2 d) and long-term (4 weeks) recovery. Seedlings were grown under controlled conditions, and irrigation was withheld until stomata closed and xylem water potential declined to -3.2 MPa on average, causing a 46% loss of stem hydraulic conductivity. Following drought release, we found a gradual recovery of leaf gas exchange to 50% to 60% of control values. This partial recovery indicates hydraulic limitations due to drought-induced damage. Whereas xylem water potential recovered close to control values within 2 d, both x-ray microtomography and intrusive measurements revealed no recovery of stem hydraulic conductivity. Moreover, we did not find indications for nonstructural carbohydrate reserves limiting hydraulic recovery. Our findings demonstrate that Scots pine is able to survive severe drought and to partially recover, although we assume that xylem development during the next growing season might compensate for some of the hydraulic impairment. Such drought-induced legacy effects are important when considering vegetation responses to extreme events.
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Roche
Amyloglucosidase, from Aspergillus niger